Stampa…

Anti-UBA52 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU1000-03LEA 909 CHF
USBIU1000-03L
Anti-UBA52 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: UBA52 (ubiquitin A-52 residue ribosomal protein fusion product 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR