Stampa…

Anti-UB Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU0999-05B-50EA 526 CHF
USBIU0999-05B-50
Anti-UB Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000-1:1500

Type: Primary
Antigen: UB (ubiquitin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Dog, Drosophila, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep, Xenopus laevis, Yeast
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR