Stampa…

Anti-UNC5C Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fornitore: United States Biological
USBIU0893-50A-APEA 1200 CHF
USBIU0893-50A-AP
Anti-UNC5C Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: UNC5C (unc-5 homolog C (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR