Stampa…
Anti-UBE2G2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: United States Biological
USBIU0880-10-PEEA 1200 CHF
USBIU0880-10-PE
Anti-UBE2G2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: UBE2G2 (ubiquitin-conjugating enzyme E2G 2)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR