Stampa…

Anti-UBE2I Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIU0880-03D-BIOTEA 1200 CHF
USBIU0880-03D-BIOT
Anti-UBE2I Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: UBE2I (ubiquitin-conjugating enzyme E2I)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR