Stampa…

Anti-UBASH3A Goat Polyclonal Antibody

Fornitore: United States Biological
USBIU0846-01DEA 774 CHF
USBIU0846-01D
Anti-UBASH3A Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:64000; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: UBASH3A (ubiquitin associated and SH3 domain containing A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR