Stampa…

Anti-ZBTB32 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT9950-50BEA 774 CHF
USBIT9950-50B
Anti-ZBTB32 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:8000; Western Blot: 2-5 ?g/ml

Type: Primary
Antigen: ZBTB32 (zinc finger and BTB domain containing 32)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR