Stampa…

Anti-TWIST1 Sheep Polyclonal Antibody (Carboxyfluorescein)

Fornitore: United States Biological
USBIT9300-02BEA 756 CHF
USBIT9300-02B
Anti-TWIST1 Sheep Polyclonal Antibody (Carboxyfluorescein)
Anticorpi
Type: Primary
Antigen: TWIST1 (twist family bHLH transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR