Stampa…

Anti-TH Chicken Polyclonal Antibody

Fornitore: United States Biological
USBIT9237-12AEA 996 CHF
USBIT9237-12A
Anti-TH Chicken Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR