Stampa…

Anti-TH Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT9237-10PEA 533 CHF
USBIT9237-10P
Anti-TH Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunofluorescence:1:100-1:200

Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR