Stampa…

Anti-TYK2 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT9220-01CEA 549 CHF
USBIT9220-01C
Anti-TYK2 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: TYK2 (tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR