Stampa…

Anti-TNFRSF12A Mouse Monoclonal Antibody [clone: 9i122]

Fornitore: United States Biological
USBIT9185-05AEA 398 CHF
USBIT9185-05A
Anti-TNFRSF12A Mouse Monoclonal Antibody [clone: 9i122]
Anticorpi
Recommended dilution: Flow Cytometry: 1:25-1:200

Type: Primary
Antigen: TNFRSF12A (tumor necrosis factor receptor superfamily, member 12A)
Clonality: monoclonal
Clone: 9i122
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR