Stampa…

Anti-TSSC1 Chicken Polyclonal Antibody

Fornitore: United States Biological
USBIT9167-02EA 667 CHF
USBIT9167-02
Anti-TSSC1 Chicken Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: TSSC1 (tumor suppressing subtransferable candidate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR