Stampa…

Anti-TRAF3 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT9161-56EA 607 CHF
USBIT9161-56
Anti-TRAF3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 1:500-1:1000; Western Blot: 1:1000

Type: Primary
Antigen: TRAF3 (TNF receptor-associated factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR