Stampa…

Anti-TNFRSF1B Mouse Monoclonal Antibody (Biotin) [clone: 6A665]

Fornitore: United States Biological
USBIT9161-26EEA 802 CHF
USBIT9161-26E
Anti-TNFRSF1B Mouse Monoclonal Antibody (Biotin) [clone: 6A665]
Anticorpi
Recommended dilution: Immunochemistry: 1:10; Flow Cytometry: 1:10; Western Blot: 1:10

Type: Primary
Antigen: TNFRSF1B (tumor necrosis factor receptor superfamily, member 1B)
Clonality: monoclonal
Clone: 6A665
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR