Stampa…

Anti-TNFRSF1A Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: 13A86]

Fornitore: United States Biological
USBIT9161-19EEA 1040 CHF
USBIT9161-19E
Anti-TNFRSF1A Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: 13A86]
Anticorpi
Recommended dilution: Flow Cytometry: 1:50

Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: monoclonal
Clone: 13A86
Conjugation: FITC (Fluorescein)
Epitope:
Host: Hamster
Isotype: IgG1
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR