Stampa…

Anti-TNFRSF1A Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT9161-07NEA 1010 CHF
USBIT9161-07N
Anti-TNFRSF1A Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR