Stampa…

Anti-TNFRSF1A Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT9161-07KEA 676 CHF
USBIT9161-07K
Anti-TNFRSF1A Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.1-0.2?g/ml; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR