Stampa…

Anti-TNFRSF1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: United States Biological
USBIT9161-07H-HRPEA 1200 CHF
USBIT9161-07H-HRP
Anti-TNFRSF1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR