Stampa…

Anti-TNFRSF1A Mouse Monoclonal Antibody [clone: MR1-2]

Fornitore: United States Biological
USBIT9161-05WEA 893 CHF
USBIT9161-05W
Anti-TNFRSF1A Mouse Monoclonal Antibody [clone: MR1-2]
Anticorpi
Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: monoclonal
Clone: MR1-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR