Stampa…
Anti-TNFRSF1A Mouse Monoclonal Antibody [clone: 3H3103]
Fornitore: United States Biological
USBIT9161-05NEA 788 CHF
USBIT9161-05N
Anti-TNFRSF1A Mouse Monoclonal Antibody [clone: 3H3103]
Anticorpi
Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: monoclonal
Clone: 3H3103
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Mehr About VWR