Stampa…

Anti-TNFAIP3 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT9160-38GEA 889 CHF
USBIT9160-38G
Anti-TNFAIP3 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: TNFAIP3 (tumor necrosis factor, alpha-induced protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR