Stampa…

Anti-TNF Mouse Monoclonal Antibody [clone: tnfa9.9]

Fornitore: United States Biological
USBIT9160-29E-100EA 733 CHF
USBIT9160-29E-100
Anti-TNF Mouse Monoclonal Antibody [clone: tnfa9.9]
Anticorpi
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: tnfa9.9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR