Stampa…

Anti-TNF Rat Monoclonal Antibody [clone: 9G292]

Fornitore: United States Biological
USBIT9160-28DEA 993 CHF
USBIT9160-28D
Anti-TNF Rat Monoclonal Antibody [clone: 9G292]
Anticorpi
Recommended dilution: Flow Cytometry: 1:50

Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: 9G292
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgD
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR