Stampa…
Anti-TNF Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT9160-17AEA 881 CHF
USBIT9160-17A
Anti-TNF Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Neutralization: 0. 012-0. 014?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR