Stampa…

Anti-TNF Mouse Monoclonal Antibody (Biotin) [clone: 6i8]

Fornitore: United States Biological
USBIT9160-04C1EA 938 CHF
USBIT9160-04C1
Anti-TNF Mouse Monoclonal Antibody (Biotin) [clone: 6i8]
Anticorpi
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: 6i8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR