Stampa…

Anti-TNF Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT9160-03UEA 1540 CHF
USBIT9160-03U
Anti-TNF Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Flow Cytometry: 1:10; Western Blot: 1:10; Immunochemistry: 1:10

Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR