Stampa…

Anti-TNF Sheep Polyclonal Antibody

Fornitore: United States Biological
USBIT9160-02MEA 988 CHF
USBIT9160-02M
Anti-TNF Sheep Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR