Stampa…

Anti-TTLL4 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT9156-76EA 774 CHF
USBIT9156-76
Anti-TTLL4 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1: 32000

Type: Primary
Antigen: TTLL4 (tubulin tyrosine ligase-like family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR