Stampa…

Anti-TUBG1 Mouse Monoclonal Antibody

Fornitore: United States Biological
USBIT9156-40NEA 465 CHF
USBIT9156-40N
Anti-TUBG1 Mouse Monoclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TUBG1 (tubulin, gamma 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR