Stampa…

Anti-TUBB Rabbit Monoclonal Antibody (Alexa Fluor® 555)) [clone: 7H79]

Fornitore: United States Biological
USBIT9155-15EA 760 CHF
USBIT9155-15
Anti-TUBB Rabbit Monoclonal Antibody (Alexa Fluor® 555)) [clone: 7H79]
Anticorpi
Recommended dilution: Immunofluorescence: 1:200
Do not freeze!

Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: monoclonal
Clone: 7H79
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila, Human, Monkey, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR