Stampa…
Anti-TUBB3 Rabbit Monoclonal Antibody [clone: 11C1086]
Fornitore: United States Biological
USBIT9155-03MEA 836 CHF
USBIT9155-03M
Anti-TUBB3 Rabbit Monoclonal Antibody [clone: 11C1086]
Anticorpi
Recommended dilution: Immunofluorescence: 1:200; Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: TUBB3 (tubulin, beta 3 class III)
Clonality: monoclonal
Clone: 11C1086
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Mehr About VWR