Stampa…

Anti-TUBB2A Mouse Monoclonal Antibody [clone: 2B2]

Fornitore: United States Biological
USBIT9155-02TEA 988 CHF
USBIT9155-02T
Anti-TUBB2A Mouse Monoclonal Antibody [clone: 2B2]
Anticorpi
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TUBB2A (tubulin, beta 2A class IIa)
Clonality: monoclonal
Clone: 2B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR