Stampa…

Anti-TUBB Mouse Monoclonal Antibody [clone: 11H5]

Fornitore: United States Biological
USBIT9155-01WEA 506 CHF
USBIT9155-01W
Anti-TUBB Mouse Monoclonal Antibody [clone: 11H5]
Anticorpi
Recommended dilution: Immunofluorescence: 1:1000-1:5000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: monoclonal
Clone: 11H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR