Stampa…

Anti-TTK Mouse Monoclonal Antibody [clone: 10F776]

Fornitore: United States Biological
USBIT9142-01BEA 687 CHF
USBIT9142-01B
Anti-TTK Mouse Monoclonal Antibody [clone: 10F776]
Anticorpi
Recommended dilution: Western Blot:1:2000-1:10000

Type: Primary
Antigen: TTK (TTK protein kinase)
Clonality: monoclonal
Clone: 10F776
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR