Stampa…

Anti-TSG101 Mouse Monoclonal Antibody [clone: 5B7]

Fornitore: United States Biological
USBIT9101-75DEA 988 CHF
USBIT9101-75D
Anti-TSG101 Mouse Monoclonal Antibody [clone: 5B7]
Anticorpi
Recommended dilution: Western Blot: 1:500-1:1000; Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TSG101 (tumor susceptibility 101)
Clonality: monoclonal
Clone: 5B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR