Stampa…

Anti-TSG101 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornitore: United States Biological
USBIT9101-75B-FITCEA 1200 CHF
USBIT9101-75B-FITC
Anti-TSG101 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: TSG101 (tumor susceptibility 101)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR