Stampa…

Anti-TSC2 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT9100-25LEA 533 CHF
USBIT9100-25L
Anti-TSC2 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA:1:4000; Western Blot:1:500-1:1000; Immunochemistry:1:50-1:100

Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR