Stampa…

Anti-TSC2 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT9100-25G6EA 825 CHF
USBIT9100-25G6
Anti-TSC2 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:500

Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR