Stampa…

Anti-TSC2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: United States Biological
USBIT9100-25G2-APCEA 1200 CHF
USBIT9100-25G2-APC
Anti-TSC2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR