Stampa…

Anti-Tryptase, epsilon Rat Monoclonal Antibody (Biotin) [clone: 9H191]

Fornitore: United States Biological
USBIT8700-20EA 1140 CHF
USBIT8700-20
Anti-Tryptase, epsilon Rat Monoclonal Antibody (Biotin) [clone: 9H191]
Anticorpi
Type: Primary
Antigen: Tryptase, epsilon
Clonality: monoclonal
Clone: 9H191
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR