Stampa…

Anti-Trypsin Mouse Monoclonal Antibody [clone: 6F201]

Fornitore: United States Biological
USBIT8669-13AEA 1200 CHF
USBIT8669-13A
Anti-Trypsin Mouse Monoclonal Antibody [clone: 6F201]
Anticorpi
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: Trypsin
Clonality: monoclonal
Clone: 6F201
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR