Stampa…

Anti-Trypsin Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT8669-05-100EA 1150 CHF
USBIT8669-05-100
Anti-Trypsin Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: Trypsin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR