Stampa…

Anti-TRPV3 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8667EA 825 CHF
USBIT8667
Anti-TRPV3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: TRPV3 (transient receptor potential cation channel, subfamily V, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR