Stampa…

Anti-TRRAP Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8666-40BEA 637 CHF
USBIT8666-40B
Anti-TRRAP Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: TRRAP (transformation/transcription domain-associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR