Stampa…

Anti-TRRAP Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT8666-40A-BIOTEA 864 CHF
USBIT8666-40A-BIOT
Anti-TRRAP Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TRRAP (transformation/transcription domain-associated protein)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR