Stampa…
Anti-TRPM8 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Fornitore: United States Biological
USBIT8666-38MEA 1450 CHF
USBIT8666-38M
Anti-TRPM8 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Anticorpi
Type: Primary
Antigen: TRPM8 (transient receptor potential cation channel, subfamily M, member 8)
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Mehr About VWR