Stampa…

Anti-Troponin T Mouse Monoclonal Antibody [clone: 10K379]

Fornitore: United States Biological
USBIT8665-23XEA 676 CHF
USBIT8665-23X
Anti-Troponin T Mouse Monoclonal Antibody [clone: 10K379]
Anticorpi
Recommended dilution: Western Blot: 2-4?g/ml; Immunochemistry: 4-8?g/ml

Type: Primary
Antigen: Troponin T
Clonality: monoclonal
Clone: 10K379
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rabbit, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR