Stampa…

Anti-TNNI3 Mouse Monoclonal Antibody [clone: 7E141]

Fornitore: United States Biological
USBIT8665-16HEA 1290 CHF
USBIT8665-16H
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 7E141]
Anticorpi
Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: monoclonal
Clone: 7E+141
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR