Stampa…

Anti-tRNA Synthetase, Alanyl Mouse Polyclonal Antibody

Fornitore: United States Biological
USBIT8663-70DEA 733 CHF
USBIT8663-70D
Anti-tRNA Synthetase, Alanyl Mouse Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: tRNA Synthetase, Alanyl
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR